Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Methyl arachidate ≥98%

Glass bottle for solids, 10 g
Catalog # 42399.09
Methyl arachidate ≥98%
Catalog # 42399.09
CAS Number:  1120-28-1
Methyl arachidate ≥98%
Methyl arachidate ≥98%

Glass bottle for solids, 10 g

CAS Number:  1120-28-1
Price
SFr. 578.00
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 98+% CAS No.: 1120-28-1 Molecular Formula: CH3(CH2)18COOCH3 Formula Weight: 326.56 Flash Point: 110°(230°F) Physical Form: Crystalline powder Melting Point: 48° Boiling Point: 215°/10mm MDL No.: MFCD00009014

Synonyms:  Methyl eicosanoate, Methyl Arachidate
  • CAS Number:1120-28-1
  • Formula:C₂₁H₄₂O₂
  • MDL Number:MFCD00009014
  • Boiling Pt:215 °C (10 mmHg)
  • Storage Temperature:Ambient
  • EINECS:214-304-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..