Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Measuring spoons

Measuring spoons

60 mm, 1,6 ml
Catalog # 216-7354
Supplier:   SEMADENI
Measuring spoons
Catalog # 216-7354
Supplier:  SEMADENI
Measuring spoons
Measuring spoons

60 mm, 1,6 ml

Price
SFr. 11.60
Log in for availability
1 * 100 ST
Product Details & Documents

Disposable measuring spoons made of transparent PS.

Specifications
  • Capacity:
    1,6 ml
  • Length:
    60 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.