Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Measuring spoons

Measuring spoons

90 mm, 10 ml
Catalog # 216-7351
Supplier:   SEMADENI
Measuring spoons
Catalog # 216-7351
Supplier:  SEMADENI
Measuring spoons
Measuring spoons

90 mm, 10 ml

Price
SFr. 23.00
Log in for availability
1 * 100 ST
Product Details & Documents

Disposable measuring spoons made of transparent PS.

Specifications
  • Capacity:
    10 ml
  • Length:
    90 mm
  • Seq:
    04