Orders LinkContactUsLinkComponent
Safety baskets
Safety baskets
Basket with handle, PP, green, 430×310×220 mm
Catalog # 216-7333
Supplier:   SEMADENI
Safety baskets
Catalog # 216-7333
Supplier:  SEMADENI
Safety baskets
Safety baskets
Basket with handle, PP, green, 430×310×220 mm
Price
38.20 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
PP, coloured.
  • Anti-static
  • Space-saving, stackable
  • Good chemical and temperature resistance
The antistatic safety baskets with perforated walls and base comes with collapsible plastic mounting bracket with lock in vertical position.
Specifications
  • Description:
    Basket with handle, PP, green
  • Seq:
    01
  • W×D×H:
    430×310×220 mm