Storage Boxes
with hinged lid and 3×6 number of compartments, Transparent, 210×120×35 mm, PS
Catalog # 216-7311
Supplier: SEMADENI
Jump to:
Product Details & Documents
These assortment and bearing boxes made of polystyrene (PS) are transparent with hinged lid and multiple longitudinal and transverse divisions.
Specifications
- Colour:Transparent
- Description:with hinged lid and 3×6 number of compartments
- Material:PS
- Seq:01
- W×D×H:210×120×35 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



