Packaging and Storage Boxes
Transparent, 95×75×33 mm, PS
Catalog # 216-3131
Supplier: SEMADENI
Jump to:
Product Details & Documents
These boxes made of polystyrene (PS) are crystal clear with a good closing tight fitting lid.
- Clear for easy product identification
- The lids are either mounted or not mounted
- Stackable
Specifications
- Colour:Transparent
- Material:PS
- Seq:15
- W×D×H:95×75×33 mm