Storage Bins
Blue, 500×315×200 mm, PP
Catalog # 215-0283
Supplier: SEMADENI
Jump to:
Product Details & Documents
Made of PP.
- Secure stacking
- Made of high quality material and are dimensionally stable
- Supporting edge and smooth inner surfaces for easy cleaning
Storage bins with large labelling field for content description.
Specifications
- Colour:Blue
- Material:PP
- W×D×H:500×315×200 mm



