ContactUsLinkComponent
SONOREX PR 140 C - PIPETTE WASHER SET, 35 KHZ, 230 V , 50/60 HZ, COMPLETE WITH PLASTIC BASKET, S/S LID AND TICKOPUR R 33 UNIVERSAL CLEANING CONCENTRATE, FOR PIPETTES UP TO 755 MM LENGTH, CLEANING AND RINSING IN THE SAME VESSEL, 13,9 L, ULTRASONIC PE 1 * 1 items
Catalog # 149-1000
undefined
SONOREX PR 140 C - PIPETTE WASHER SET, 35 KHZ, 230 V , 50/60 HZ, COMPLETE WITH PLASTIC BASKET, S/S LID AND TICKOPUR R 33 UNIVERSAL CLEANING CONCENTRATE, FOR PIPETTES UP TO 755 MM LENGTH, CLEANING AND RINSING IN THE SAME VESSEL, 13,9 L, ULTRASONIC PE 1 * 1 items
Catalog # 149-1000