ContactUsLinkComponent
SONOREX SUPER RK 31 H, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 190X85X60 MM 0,9 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEATIN G FIXED AT 65C/149F, ULTRASONIC PEAK OUTPUT 240 W, HF OUTPUT: 30 WEFF. 1 * 1 items
Catalog # 142-2521
undefined
SONOREX SUPER RK 31 H, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 190X85X60 MM 0,9 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HEATIN G FIXED AT 65C/149F, ULTRASONIC PEAK OUTPUT 240 W, HF OUTPUT: 30 WEFF. 1 * 1 items
Catalog # 142-2521
Frequently Bought Together