ContactUsLinkComponent
TICKOPUR R 30 - 5 LITRES, NEUTRAL CLEANER, CONCENTRATE, WITH CORROSION PROTECTION, GENTLE TO MATERIAL, DOSAGE 1 - 5 %, PH 7 (1 %) 1 * 5 L
Catalog # 142-1931
TICKOPUR R 30 - 5 LITRES, NEUTRAL CLEANER, CONCENTRATE, WITH CORROSION PROTECTION, GENTLE TO MATERIAL, DOSAGE 1 - 5 %, PH 7 (1 %) 1 * 5 L
Catalog # 142-1931