ContactUsLinkComponent
SONOREX SUPER RK 100 SH, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 240X140X100 MM 3,0 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HE ATING 30-80C (86-176F), OUTLET: HOSE, ULTRASONIC PEAK OUTPUT 320 W, HF-OUTPUT: 80 WEFF. 1 * 1 items
Catalog # 142-1140
SONOREX SUPER RK 100 SH, ULTRASONIC BATH, 35 KHZ, 230 V 50/60 HZ, ID 240X140X100 MM 3,0 L, S/S TIMER 1-15 MIN AND CONTINUOUS, HE ATING 30-80C (86-176F), OUTLET: HOSE, ULTRASONIC PEAK OUTPUT 320 W, HF-OUTPUT: 80 WEFF. 1 * 1 items
Catalog # 142-1140
Frequently Bought Together