Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Potassium dichromate standardised solution 0.025 N in aqueous solution

Potassium dichromate standardised solution 0.025 N in aqueous solution

Glass bottle, 500 mL
Catalog # 35589.AP
Health
Irritant
Potassium dichromate standardised solution 0.025 N in aqueous solution
Catalog # 35589.AP
CAS Number:  7778-50-9
Potassium dichromate standardised solution 0.025 N in aqueous solution
Potassium dichromate standardised solution 0.025 N in aqueous solution

Glass bottle, 500 mL

CAS Number:  7778-50-9
Price
SFr. 50.60
Log in for availability
1 * 500 mL
Product Details & Documents

CAS No.: 7778-50-9 Molecular Formula: Cr2K2O7 Formula Weight: 294.18 Physical Form: Liquid MDL No.: MFCD00011367

  • CAS Number:7778-50-9
  • Formula:K₂Cr₂O₇
  • MDL Number:MFCD00011367
  • Storage Temperature:Ambient
  • EINECS:231-906-6
  • UN:3287
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.