Laboratory bottles, narrow neck, with thread
1000 ml, GL 45, 230 mm, 95×95×230 mm, Amber
Catalog # 215-3838
Supplier: Brand
Jump to:
Product Details & Documents
Soda-lime glass, amber, coated or uncoated.
- Good chemical and physical properties
- Maximum operating temperature for coated bottles is 80 °C
- To preserve the coating, do not clean at temperatures exceeding 60 °C
DIN thread. Space saving in storage and transport. Stable, non-slip. Amber to protect light-sensitive materials. The coating envelops the glass bottle like a protection skin. Should the bottle break, the splintering hazards are reduced considerably.
Documents
Brand Dispensette® S
Product Literature
BRND704010_TDS_EN BRND704020_TDS_EN
Datasheet
GA Dispensette S
Manual
Specifications
- Thread:GL 45
- Capacity:1000 ml
- Colour:Amber
- Height:230 mm
- Seq:04
- W×D×H:95×95×230 mm



