Plating beads, ColiRollers™
Plating beads
Supplier: Merck Millipore (Novagen)
Jump to:
Product Details & Documents
ColiRollers™ plating beads are a specially treated glass bead which offers a better way to perform plating bacteria for transformation and molecular cloning applications. ColiRollers™ beads eliminate the inconsistencies of conventional 'hockey stick' methods.
- Non laborious and time saving
- Entire stacks of plates can be processed at one time
- Cells are evenly and completely spread over the agar surface
Delivery information: Each pack contains enough ColiRollers™ beads for 60 to 80 plates.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Plating beads
- Seq:02