Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Mortars, without pestles

Mortars, without pestles

500 ml, 90 mm, 150 mm
Catalog # 410-0084
Supplier:   VITLAB
Mortars, without pestles
Catalog # 410-0084
Supplier:  VITLAB
Mortars, without pestles
Mortars, without pestles

500 ml, 90 mm, 150 mm

Price
44.20 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

White Melamine, with lip.

 : Pestle must be ordered separately.

Specifications
  • Ø:
    150 mm
  • Capacity:
    500 ml
  • Height:
    90 mm