Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Mortars, without pestles

Mortars, without pestles

300 ml, 75 mm, 125 mm
Catalog # 410-0083
Supplier:   VITLAB
Mortars, without pestles
Catalog # 410-0083
Supplier:  VITLAB
Mortars, without pestles
Mortars, without pestles

300 ml, 75 mm, 125 mm

Price
SFr. 36.10
Log in for availability
1 * 1 ST
Product Details & Documents

White Melamine, with lip.

Ordering information: Pestle must be ordered separately.

Specifications
  • Ø:
    125 mm
  • Capacity:
    300 ml
  • Height:
    75 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.