Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tripods

Tripods

With detachable legs, 160×210 mm
Catalog # 451-9109
Supplier:   RSG
Tripods
Catalog # 451-9109
Supplier:  RSG
Tripods
Tripods

With detachable legs, 160×210 mm

Price
SFr. 58.70
Log in for availability
1 * 1 ST
Product Details & Documents

Separable tripods made from stainless steel.

Specifications
  • Int.Ø×H:
    160×210 mm
  • Description:
    With detachable legs
  • Seq:
    04
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Tripod extensions, set of three, total height up to 370 mm
    Tripod extensions, set of three, total height up to 370 mm
    RSGA497.370
    Log in for availability
    1 * 3 ST - 84.60 SFr.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.