Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Tripods

With detachable legs, 100×210 mm
  451-9107
 :   RSG
Tripods
  451-9107
 :  RSG
Tripods
Tripods

With detachable legs, 100×210 mm

 
SFr. 54.60
   
1 * 1 ST
 

Separable tripods made from stainless steel.

 
  • Int.Ø×H:
    100×210 mm
  • Description:
    With detachable legs
  • Seq:
    01
Accessories

 

  •  
    Description
     
     
  • RSGA497.370
    Tripod extensions, set of three, total height up to 370 mm
       
    1 * 3 ST - SFr. 84.60