Support rings
Heavy support ring, with rod, open, with rubber coating, 100 mm
RSGA283.100
: RSG
These heavy-duty support rings are made from stainless steel.
- 8 mm rod
- Available with rubber covered rings
- Ø int.:100 mm
- Description:Heavy support ring, with rod, open, with rubber coating
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



