Support rings
Heavy support ring, with rod (8 mm), 130 mm
Catalog # 241-0319
Supplier: RSG
Jump to:
Product Details & Documents
These heavy-duty support rings are made from stainless steel.
- 8 mm rod
- Available with rubber covered rings
Specifications
- Ø int.:130 mm
- Description:Heavy support ring, with rod (8 mm)
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



