Support rings
Heavy support ring, with rod (8 mm), 100 mm
RSGA280.100
: RSG
These heavy-duty support rings are made from stainless steel.
- 8 mm rod
- Available with rubber covered rings
- Ø int.:100 mm
- Description:Heavy support ring, with rod (8 mm)



