Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Support rings

Support rings

Heavy support ring, with rod (8 mm), 70 mm
Catalog # 241-0318
Supplier:   RSG
Support rings
Catalog # 241-0318
Supplier:  RSG
Support rings
Support rings

Heavy support ring, with rod (8 mm), 70 mm

Price
32.10 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

These heavy-duty support rings are made from stainless steel.

  • 8 mm rod
  • Available with rubber covered rings
Specifications
  • Ø int.:
    70 mm
  • Description:
    Heavy support ring, with rod (8 mm)