Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Support rings

Support rings

Heavy support ring, with rod (8 mm), 50 mm
Catalog # 241-0317
Supplier:   RSG
Support rings
Catalog # 241-0317
Supplier:  RSG
Support rings
Support rings

Heavy support ring, with rod (8 mm), 50 mm

Price
SFr. 29.10
Log in for availability
1 * 1 ST
Product Details & Documents

These heavy-duty support rings are made from stainless steel.

  • 8 mm rod
  • Available with rubber covered rings
Specifications
  • Ø int.:
    50 mm
  • Description:
    Heavy support ring, with rod (8 mm)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.