Microscopy instrument set
Microscopy set, 5 pcs.
Catalog # RSGA066.805
Supplier: RSG
Jump to:
Product Details & Documents
Microscopy instrument set in a grey wooden box, all articles are completely made of stainless steel.
- 1 pair of microscopy scissors
- 1 microscopy needle straight
- 1 microscopy needle bent
- 1 pair of tweezers, pointed straight with guide pin
- 1 scalpel
Ordering information: On demand the arrangement of the 7 pcs. set can be individualised.
Specifications
- Description:Microscopy set, 5 pcs.