Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Accessories for empty columns HR series

Accessories for empty columns HR series

Preflanged tubing, 400 mm, HR 16, I.Ø 0,5 mm, O.Ø 1,8 mm
Catalog # 18-0421-01
Supplier:   Cytiva
Accessories for empty columns HR series
Catalog # 18-0421-01
Supplier:  Cytiva
Accessories for empty columns HR series
Accessories for empty columns HR series

Preflanged tubing, 400 mm, HR 16, I.Ø 0,5 mm, O.Ø 1,8 mm

Price
SFr. 116.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Preflanged tubing, 400 mm, HR 16, I.Ø 0,5 mm, O.Ø 1,8 mm
  • Seq:
    11
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.