ContactUsLinkComponent
Superdex Peptide PC 3.2/30 Gel Filtration Media pre-packed in PC Column for SMART System (PC columns can be used in other systems th an the SMART System by using the PC Holder 17-1455-01) 1 * 1 items
Catalog # 17-1458-01
Supplier:  Cytiva
Superdex Peptide PC 3.2/30 Gel Filtration Media pre-packed in PC Column for SMART System (PC columns can be used in other systems th an the SMART System by using the PC Holder 17-1455-01) 1 * 1 items
Catalog # 17-1458-01
Supplier:  Cytiva
Frequently Bought Together