Orders LinkContactUsLinkComponent
Diamond pen
Diamond pen, wooden shaft
  201-0391
Diamond pen
  201-0391
Diamond pen
Diamond pen
Diamond pen, wooden shaft
SFr. 49.50
1 * 1 ST
Diamond tip in metal holder.
  • Available with wooden handle or in the form of a ballpoint pen
  • Not suitable for engraving
  • 140 mm long
For labelling glass and metal surfaces.
  • Description:
    Diamond pen, wooden shaft