Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Stand bases

Stand bases

300×150 mm, 3,7 kg
Catalog # USBE2014-2144
Supplier:   USBECK Laborgeräte
Stand bases
Catalog # USBE2014-2144
Stand bases
Stand bases

300×150 mm, 3,7 kg

Price
SFr. 186.00
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel. Firm, non slip stand for a wide variety of uses. With an M10 threaded hole on face side and adjustable feet for uneven surfaces.

  • Rubber feet for a firm footing
  • Smooth surfaces for easy, fast cleaning
  • Robust

DIN 12892/55

Certifications: DIN 12892/55

Documents

Specifications
  • L×W:
    300×150 mm
  • Seq:
    04
  • Weight:
    3,7 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.