Screw Top Vial Clear Autosystem(Pkg 100) (PERKIN N9302945) 1 * 100 items
Catalog # PERCN9302945
Supplier: PerkinElmer
Screw Top Vial Clear Autosystem(Pkg 100) (PERKIN N9302945) 1 * 100 items
Catalog # PERCN9302945
Supplier: PerkinElmer
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR® Safety Wash Bottles
Supplier: VWR Collection
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Shell Vials
Supplier: VWR Collection