Orders LinkContactUsLinkComponent
Closures for headspace vials | Avantor
Closures for headspace vials
Red PTFE coated butyl rubber cap, pre-assembled
Catalog # PERCB4000025
Supplier:   PerkinElmer
Closures for headspace vials
Catalog # PERCB4000025
Supplier:  PerkinElmer
Closures for headspace vials
Red PTFE coated butyl rubber cap, pre-assembled
Price
SFr. 1’520.00
Log in for availability
1 * 1.000 ST
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Specifications
  • Description:
    Red PTFE coated butyl rubber cap, pre-assembled
  • Seq:
    07