Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Probes for temperature and humidity data loggers

Probes for temperature and humidity data loggers

Probe, needle, T-type, 1,25 ml and 12 g, Length: 1340 mm, probe shaft Ø×L: 1,4×60 mm, −50…+250 °C, T: ±0.2 °C (–20...+70 °C); Class 1: (–40...350 °C)
Catalog # 620-3319
Supplier:   Testo
Probes for temperature and humidity data loggers
Catalog # 620-3319
Supplier:  Testo
Probes for temperature and humidity data loggers
Probes for temperature and humidity data loggers

Probe, needle, T-type, 1,25 ml and 12 g, Length: 1340 mm, probe shaft Ø×L: 1,4×60 mm, −50…+250 °C, T: ±0.2 °C (–20...+70 °C); Class 1: (–40...350 °C)

Price
SFr. 138.00
Log in for availability
1 * 1 ST
Product Details & Documents

Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature. Humidity sensor is durable and traceable to international humidity standards, such as ILAC, PTB and NIST. Thus, even after years of use you can still count on measurement results that are reliable and correct.

Specifications
  • Range:
    −50…+250 °C
  • Description:
    Probe, needle, T-type, 1,25 ml and 12 g, Length: 1340 mm, probe shaft Ø×L: 1,4×60 mm
  • Seq:
    09
  • Accuracy:
    T: ±0.2 °C (–20...+70 °C); Class 1: (–40...350 °C)