Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Ion exchangers

Strong acid cation exchanger, Amberlite® IR-120, H+, exchange capacity >1,7 mval/ml, 500 mL
Catalog # 1.15131.0500
Supplier:   Merck
Ion exchangers
Catalog # 1.15131.0500
Supplier:  Merck
CAS Number:  
Ion exchangers

Strong acid cation exchanger, Amberlite® IR-120, H+, exchange capacity >1,7 mval/ml, 500 mL

CAS Number:  
Price
SFr. 77.50
Log in for availability
1 * 500 mL
Product Details & Documents

In general, ion exchangers consist of a cross-linked polymer matrix with a uniform distribution of fixed ionic groups sited throughout the resin structure. These must be balanced by a similar number of ions of the opposite charge, (counter ions) to maintain electrical neutrality. Cation exchangers therefore, exchange and enrich only cations, anion exchangers only anions. In contrast adsorber resins have a non ionic, but depending on the structure, a somewhat polar character, and don't adsorb stoichiometrically both: anions, cations, as well as uncharged compounds.

Possible applications are:
Trace enrichment by using chelating ion exchangers
Determination of total salt content of solutions and water by H+ exchange
Removals of interfering cations or anions
Chromatographic separation
Digestion of insoluble compounds
Application as a catalyst

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Strong acid cation exchanger, Amberlite® IR-120, H<sup>+</sup>, exchange capacity >1,7 mval/ml

Specifications