Jump to:
Product Details & Documents
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
- Roll format are easy to use and most common ranges are available in a refill format
- Quality assurance - tests are checked and calibrated against certified buffer solutions
- Wide and narrow pH ranges available
* no colour scale card supplied
** pack of 3×100 strips
Specifications
- pH gradation:1
- pH range:-
- Description:Replacement rolls for pH 1 - 14*
- Cat. No.:1.10232.0001
- M-Clarity quality level:MQ100