Buffer solutions, Certipur®, colour coded, Supelco®
Catalog # 1.09476.9010
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Materials for the precise calibration and monitoring of pH meters.
- Traceable to NIST primary standard and PTB - Uncertainty ±0,015 (except pH 10 with ±0,05)
- Colour coded solutions for quick identification
- Larger pack sizes available, the easy to use Titripac® packaging eliminates contamination issues
- Reference temperature: +20 °C
Measured in an independent accredited calibration laboratory for the pH value measurement (DIN EN ISO/IEC 17025).
Documents
Certificates
Search by Lot NumberCertipur Merck Buffer Calibration Standards for pH Measurements
Product Literature
ProductPage_109475 ProductPage_109476 ProductPage_109477 Spec MERC1.09400.0500.pdf
Product specification
Specifications
Specifications
- Pack type:Titripac®
- pH buffer type:Borate
- Pk:10 L
- Description:Certipur® buffer solution, pH 9.00, blue
- Cat. No.:1.09476.9010
- M-Clarity quality level:MQ300
- Environmentally Preferable:Sustainable Packaging



