Buffer solutions, Certipur®, colour coded, Supelco®
Certipur® buffer solution, pH 4,00, red, Citrate, Plastic bottle, 500 mL
Anbieter: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Katalognr. 1.09475.0500
Anbieter: Merck
CAS Nummer:
Direkt zu
Product Details & Documents
Materials for the precise calibration and monitoring of pH meters.
- Traceable to NIST primary standard and PTB - Uncertainty ±0,015 (except pH 10 with ±0,05)
- Colour coded solutions for quick identification
- Larger pack sizes available, the easy to use Titripac® packaging eliminates contamination issues
- Reference temperature: +20 °C
Measured in an independent accredited calibration laboratory for the pH value measurement (DIN EN ISO/IEC 17025).
Documents
Zertifikate
Suche nach ChargennummerCertipur Merck Buffer Calibration Standards for pH Measurements
Product Literature
ProductPage_109475 ProductPage_109476 ProductPage_109477 Spec MERC1.09400.0500.pdf
Product specification
Technische Daten
Technische Daten
- Pack type:Plastic bottle
- pH buffer type:Citrate
- Pk:500 mL
- Description:Certipur® buffer solution, pH 4,00, red
- Cat. No.:1.09475.0500
- M-Clarity quality level:MQ300



