Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Sorbents, LiChrosorb®

LiChrosorb® RP-8, 10 µm, 10 g
Catalog # 1.09318.0010
Supplier:   Merck
Sorbents, LiChrosorb®
Catalog # 1.09318.0010
Supplier:  Merck
CAS Number:  
Sorbents, LiChrosorb®
Sorbents, LiChrosorb®

LiChrosorb® RP-8, 10 µm, 10 g

CAS Number:  
Price
1 320.00 SFr.
Log in for availability
1 * 10 g
Product Details & Documents

LiChrosorb® is an irregular shaped silica sorbent. LiChrosorb® packing materials offer the compete programme of non-polar derivatives (RP-8, RP-18, RP-select B) and polar derivatives (Si 60 and Si 100).

Specifications

Specifications

  • Particle Size:
    10 µm
  • Pk:
    10 g
  • Description:
    LiChrosorb® RP-8

Specifications