Potassium hydroxide, pellets, EMSURE® for analysis, Supelco®
Fibre carton, 50 kg
Supplier: Merck
Potassium hydroxide, pellets, EMSURE® for analysis, Supelco®
Catalog # 1.05033.9050
Supplier: Merck
CAS Number: 1310-58-3
Jump to:
Product Details & Documents
Synonyms: Potash caustic, Caustic potash
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Merck Index:12,07806
- Boiling Pt:1320 °C (1013 hPa)
- Melting Pt:360 °C
- Density:2.04 g/cm³ (20 °C)
- EINECS:215-181-3
- ADR:8,II
- :M-Clarity™ quality level = MQ300
- Storage Temperature:Ambient
- UN:1813
- REACH:01-2119487136-33
Documents
Certificates
Search by Lot NumberMERC1.05033.0500_TDS MERC1.05033.1000_TDS_EN ProductPage_105033
Product specification
Merck Individual products and tailor-made chemicals for your technical applications
Product Literature
msds_105033
SDS
Specifications
Specification Test Results
- Solubility:1130 g/l (20 °C)
- Melting Point:360 °C
- Molar Mass:56.11 g/mol
- Boiling Point:1320 °C
- Vapor Pressure:(20 °C)
- Density:2.04 g/cm3 (20 °C)
- pH:14 (56 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specifications
- Pack type:Fibre carton
- Pk:50 kg



