Orders LinkContactUsLinkComponent
Scoops
Scoops
500 ml, 325 mm
Catalog # NICK9030
Supplier:   NICKEL ELECTRO
Scoops
Catalog # NICK9030
Supplier:  NICKEL ELECTRO
Scoops
Scoops
500 ml, 325 mm
Price
SFr. 52.80
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel scoops for collecting or measuring powders, pastes and granulates.
  • Excellent thermal resistance, autoclavable
  • Non magnetic, bright finish
  • Highly polished surface makes cleaning easier
Specifications
  • Capacity:
    500 ml
  • Length:
    325 mm