Jump to:
Product Details & Documents
Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
- CAS Number:1314-62-1
- Formula:O₅V₂
- MDL Number:MFCD00011457
- Molecular Weight:181.88 g/mol
- Merck Index:13,09987
- Boiling Pt:1750 °C (1013 hPa)
- Melting Pt:690 °C
- Density:3.35 g/cm³ (20 °C)
- EINECS:215-239-8
- ADR:6.1,III
- :M-Clarity™ quality level = MQ200
- UN:2862
Specifications
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Melting Point:690 °C
- Molar Mass:181.88 g/mol
- Bulk Density:400 - 600 kg/m3
- Boiling Point:1750 °C (decomposition)
- Density:3.36 g/cm3 (20 °C)
- pH:4 (50 g/l, H2O, 20 °C) (slurry)
Specifications
- Pack type:Plastic bottle
- Pk:1 kg



