Cavity slides
Cavity slides with epoxy coating, White, 12, 5 mm
Catalog # 631-0452
Supplier: Epredia
Jump to:
Product Details & Documents
These autoclavable slides are specially designed for immunofluorescence.
- Adhesive surface and hydrophobic mask
- Hydrophobic epoxy- or PTFE-coating
- (*) with rings or wells
Developped for clinical diagnosis applications.
Ordering information: Available with rings or wells (*).
Documents
Specifications
- No. of wells:12
- Colour:White
- Description:Cavity slides with epoxy coating
- Seq:011
- Well Ø:5 mm