Orders LinkContactUsLinkComponent
Nalgene™ Screw Caps
Nalgene™ Screw Caps
Replacement closure, 53 B, Amber, PP
Katalognr. NALG71-2171-0530
   Low Manufacturing Impact
Nalgene™ Screw Caps
Katalognr. NALG71-2171-0530
   Low Manufacturing Impact
Nalgene™ Screw Caps
Nalgene™ Screw Caps
Replacement closure, 53 B, Amber, PP
Preis
24.10 SFr.
Einloggen für Verfügbarkeit
1 * 12 ST
Product Details & Documents
PP, coloured.
  • Very good chemical resistance
  • Heat resistant up to 135 °C
  • Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Technische Daten
  • Cap Size:
    53 B
  • Colour:
    Amber
  • Description:
    Replacement closure
  • Material:
    PP
  • Seq:
    46
  • Cat. No.:
    NALG71-2171-0530
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact