Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Buffer solutions

Type L4698, 4,00/7,00/10,01, ±0,010, 25 °C, 60 Ampoul
  662-6180
Buffer solutions
  662-6180
 :  
Buffer solutions
Buffer solutions

Type L4698, 4,00/7,00/10,01, ±0,010, 25 °C, 60 Ampoul

 :  
 
305.00 SFr.
   
1 * 60 Ampull
 

Buffer solutions intended for extremely precise calibration.

  • Highest measurement reliability
  • No preservative agents
  • Hermetically sealed in the glass ampoule
  • Sterilised with hot steam

 : According to DIN 19 266.

 

 

  • Pk:
    60 Ampoul
  • Reference temperature:
    25 °C
  • Description:
    Type L4698
  • pH accuracy:
    ±0,010
  • pH value:
    4,00/7,00/10,01

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.