Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Reference electrodes

Reference electrodes with free end, Ag/AgCl reference system and refilling tube, 9828/95, +5...+135 °C, Glass, KCI 3 mol/l, Ceramic, 12×120 mm, 5 m fixed cable
Catalog # SCOI285116851
Supplier:   SI Analytics
Reference electrodes
Catalog # SCOI285116851
Supplier:  SI Analytics
Reference electrodes
Reference electrodes

Reference electrodes with free end, Ag/AgCl reference system and refilling tube, 9828/95, +5...+135 °C, Glass, KCI 3 mol/l, Ceramic, 12×120 mm, 5 m fixed cable

Price
SFr. 432.00
Log in for availability
1 * 1 ST
Product Details & Documents

These reference electrodes are available with low maintenance plug heads or with fixed cables and free end.

Specifications
  • Connection:
    5 m fixed cable
  • Ø×L:
    12×120 mm
  • Type:
    9828/95
  • Description:
    Reference electrodes with free end, Ag/AgCl reference system and refilling tube
  • Material:
    Glass
  • Diaphr.:
    Ceramic
  • Electrolyte:
    KCI 3 mol/l
  • Temp.:
    +5...+135 °C