Orders LinkContactUsLinkComponent
Reference electrodes
Reference electrodes
Reference electrode, low maintenance, Ag/AgCl reference system, B 1881 HD, −5...+100 °C, PVDF, Paste, PVDF, 12×120 mm, Screw plug head
Catalog # SCOI285111767
Supplier:   SI Analytics
Reference electrodes
Catalog # SCOI285111767
Supplier:  SI Analytics
Reference electrodes
Reference electrodes
Reference electrode, low maintenance, Ag/AgCl reference system, B 1881 HD, −5...+100 °C, PVDF, Paste, PVDF, 12×120 mm, Screw plug head
Price
SFr. 408.00
Log in for availability
1 * 1 ST
Product Details & Documents
These reference electrodes are available with low maintenance plug heads or with fixed cables and free end.
Specifications
  • Connection:
    Screw plug head
  • Ø×L:
    12×120 mm
  • Type:
    B 1881 HD
  • Description:
    Reference electrode, low maintenance, Ag/AgCl reference system
  • Material:
    PVDF
  • Diaphr.:
    PVDF
  • Electrolyte:
    Paste
  • Temp.:
    −5...+100 °C
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.