Dropping bottles, LDPE, Nalgene®
4 ml
Catalog # 215-1063
Supplier: Thermo Fisher Scientific
Dropping bottles, LDPE, Nalgene®
Catalog # 215-1063
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
LDPE, transparent with colour-coded (red, green, blue, white and yellow) PP screw caps and LDPE dosing tip.
- Good chemical resistance, heat resistant up to 80 °C
- Drop control tip snaps securely into place, drops are dispensed individually
- Flexible for easy handling, leakproof
- Delivers 40 µl drops (based on water; viscosity affects drop size)
Reagent identification is very easy with the varied colours of the screw caps. These dropping bottles provide reliable, repeatable dispensing of reagents and other liquids. They serve as a very good alternative to pipettes or other conventional dispensing devices. They are suitable for most biotech, diagnostic and pharmaceutical applications.
Specifications
- Capacity:4 ml
- Seq:01
- Cat. No.:215-1063
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



