Dropping bottles, LDPE, Nalgene®
4 ml, LDPE, translucent
Catalog # 215-7173
Supplier: Thermo Fisher Scientific
Dropping bottles, LDPE, Nalgene®
Catalog # 215-7173
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
LDPE, translucent or LDPE, white with white PP screw cap and LDPE dosing tip.
- Good chemical resistance, heat resistant up to +80 °C
- Drop-control tip snaps securely into place, drops are dispensed individually
- Flexible for easy handling, leakproof
- Delivers 40 µl drops (based on water; viscosity affects drop size)
These dropping bottles provide reliable, repeatable dispensing of reagents and other liquids. They serve as a very good alternative to pipettes or other conventional dispensing devices. They are suitable for most biotech, diagnostic and pharmaceutical applications. White material, ideal for UV light-sensitive products.
Specifications
- Capacity:4 ml
- Material:LDPE, translucent
- Seq:01
- Cat. No.:215-7173
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact



