Specimen loops
Specimen loop, 0,5 mm, 1 mm, 60 mm
Catalog # MART64-064-01
Supplier: KLS Martin Group
Jump to:
Product Details & Documents
Specifications
- Ø:1 mm
- Description:Specimen loop, 0,5 mm
- Length:60 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



