Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Specimen loops

Specimen loops

Specimen loop, 0,5 mm, 3 mm, 40 mm
Catalog # 391-0135
Supplier:   KLS Martin Group
Specimen loops
Catalog # 391-0135
Supplier:  KLS Martin Group
Specimen loops

Specimen loop, 0,5 mm, 3 mm, 40 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Ø:
    3 mm
  • Description:
    Specimen loop, 0,5 mm
  • Length:
    40 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.