Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Osteotomes, Stille

Osteotomes, Stille

Osteotome, Stille, 205 mm, 15 mm
Catalog # MART23-124-15
Supplier:   KLS Martin Group
Osteotomes, Stille
Catalog # MART23-124-15
Supplier:  KLS Martin Group
Osteotomes, Stille
Osteotomes, Stille

Osteotome, Stille, 205 mm, 15 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

These osteotomes for bone surgery are made from stainless steel (18/10).

Specifications
  • Description:
    Osteotome, Stille
  • Length:
    205 mm
  • Width:
    15 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.