ContactUsLinkComponent
ProtoGel (30%) / 37.5:1 Acrylamide to Bisacrylamide Stabilized Solution Optimized for SDS-PAGE of Proteins (Laemmli gels) 1 * 1 l
Catalog # NADIEC-890-1L
ProtoGel (30%) / 37.5:1 Acrylamide to Bisacrylamide Stabilized Solution Optimized for SDS-PAGE of Proteins (Laemmli gels) 1 * 1 l
Catalog # NADIEC-890-1L
Frequently Bought Together